Learn More
Medchemexpress LLC Lesopitron dihydrochloride | 132449-89-9 | MFCD00894211 | 98.0% | 393.74 g/mol | C15H23Cl3N6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1016091MG
Encompass_Preferred
Lesopitron dihydrochloride is a selective, full agonist of the serotonin 5-HT1A receptor used for preclinical in vitro pharmacology and receptor-binding studies. It shows an IC50 of 125 nM in rat hippocampal membranes and is supplied as the dihydrochloride salt at high purity for research use.
  • High purity (98.0%).
  • Molecular formula C15H23Cl3N6; molecular weight 393.74 g/mol.
  • Selective 5-HT1A receptor agonist (IC50 = 125 nM in rat hippocampal membranes).
  • Suitable for in vitro receptor pharmacology and binding assays.
  • Available in small research pack sizes (e.g., 1 mg).
  • Analytical documentation available (COA, HNMR, SDS).
Catalog No. 50-002-49042