Learn More
Apexbio Technology LLC Lestaurtinib,1mg CAS# 111358-88-4
SDP
Supplier:  Apexbio Technology LLC A45131
Encompass_Preferred
Potent JAK2, FLT3 and TrkA inhibitor (IC50 values are 0.9, 3 and < 25 nM respectively) that prevents STAT5 phosphorylation (IC50 = 20 - 30 nM). Exhibits antiproliferative activity in vitro (IC50 = 30 - 100 nM in HEL92.1.7 cells) and is effective against myeloproliferative disorders in vivo. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0285