Learn More
Medchemexpress LLC Leu-AMS | 288591-93-5 | 99.6% | 459.48 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY10890010MM/1ML
Encompass_Preferred
Leu-AMS (compound 6), a leucine analogue, is a potent inhibitor of leucyl-tRNA synthetase (LRS) with an IC50 of 22.34 nM. It inhibits the catalytic activity of LRS but does not affect leucine-induced mTORC1 activation. Leu-AMS exhibits cytotoxicity in cancer cells and normal cells and inhibits bacterial growth.
  • Potent inhibitor of leucyl-tRNA synthetase (LRS) with an IC50 of 22.34 nM.
  • Inhibits the catalytic activity of LRS.
  • Does not affect leucine-induced mTORC1 activation.
  • Shows cytotoxicity in cancer cells and normal cells.
  • Inhibits the growth of bacteria.
Catalog No. 50-003-44619