Learn More
Bachem H-Leu-OMe · HCl
SDP
Supplier:  Bachem 4000607.0100
Encompass_Preferred
H-Leu-OMe · HCl, 7517-19-3, C7H15NO2 · HCl, Mr 181.66, 100g. Leu-OMe (LME) causes lysosomal disruption and death of human monocytes (M phi). In addition, LME removed natural killer cell (NK) activity from human peripheral mononuclear cells (PBM). During incubation of human lymphocytes with Leu-OMe, the dipeptide Leu-Leu-OMe which is responsible for the NK-toxicity, is formed.
Catalog No. 50-194-6955