Learn More
Cayman Chemical Leu144Arg147-PLP139-151 5mg
SDP
Supplier:  Cayman Chemical 368955 MG
Encompass_Preferred
A mutant fragment of PLP peptide; increases spleen levels of IL-4 in mice when administered at 50 UG emulsified in CFA as an immunization; inhibits Th1 cell activation in vitro but not in vivo, where it induces the generation of regulatory T cells; delays the onset of PLP (178-191)-, MOG (92-106)-, or MBP-induced EAE in mice
Catalog No. 50-291-5042