Learn More
Bachem (Leu31,Pro34)-Neuropeptide Y (porcine) Trifluoroacetate
SDP
Supplier:  Bachem 4018713.1000
Encompass_Preferred
(Leu31,Pro34)-Neuropeptide Y (porcine) Trifluoroacetate, H-Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Leu-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Leu-Thr-Arg-Pro-Arg-Tyr-NH2, 125580-28-1, C190H286N54O56, Mr 4222.69, 1mg. [Leu31, Pro34] Neuropeptide Y (1-36), porcine. This NPY analog is a highly potent and selective ligand for Y1 receptors in different cell lines and tissues. It has therefore been used to detect the NPY/PYY receptor subtype distribution, e.g., in rat brain. The peptide is also active in vivo, showing an even higher potency than NPY in increasing blood pressure in anesthetized rats.
Catalog No. 50-260-196