Learn More
Medchemexpress LLC Leucic acid | 498-36-2 | MFCD00004246 | 99.2% | 132.16 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY30216A10MM/1ML
Encompass_Preferred
Leucic acid, also known as α-Hydroxyisocaproic acid, is an orally active end-product of the microbial metabolism of leucine. It plays a role in various biological processes, including binding to HCAR2 and altering AMPK and ERK1/2 phosphorylation status. This compound has been noted for its ability to suppress lipid synthesis, promote catabolism, and reduce adiposity.
  • Enhances lean mass.
  • Improves exercise capacity.
  • Suppresses pro-inflammatory cytokine secretion.
  • Decreases basal protein synthesis.
  • Attenuates myotube atrophy.
  • Useful for obesity research.
Catalog No. 50-004-04373