Learn More
Medchemexpress LLC Leucine-2,3,3,4,5,5,5,5',5',5'-d10 | 29909-01-1 | 98.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1674S
Encompass_Preferred
(±)-Leucine-d10 is a deuterium labeled isomer of Leucine. It is used as a chemosterilant and dietary additive, notably inhibiting the growth of Escherichia coli HfrH.
  • Used as a tracer in various applications.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-14284