Learn More
Medchemexpress LLC Leucine dehydrogenase, Microorganism | 9082-71-7 | 43 kDa | 100 U

Supplier: Medchemexpress LLC HYP2768100U
Leucine dehydrogenase, Microorganism (EC 1.4.1.9) is purified from Bacillus spheroides. It catalyzes the oxidative deamination of various L-amino acids and the reductive amination of their keto analogues.
- Purified from Bacillus spheroides.
- Catalyzes both oxidative deamination and reductive amination of various L-amino acids and their keto analogues.
- Molecular weight of 43 kDa and an isoelectric point of 6.6.
- pH stable from 6.0-11.0 (25°C, 15 hr) and optimum temperature is 55-60°C.
- Inhibited by Hg2+.
- Specific activity of ≥25 U/mg protein.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.