Learn More
Medchemexpress LLC (±)-Leucine (DL-Leucine) | 328-39-2 | 99.95% | 50 G
SDP
Supplier:  Medchemexpress LLC HYB167450G
Encompass_Preferred
(±)-Leucine (DL-Leucine) is an isomer of Leucine that acts as a chemosterilant and dietary additive. It has been shown to inhibit the growth of *Escherichia coli* HfrH by 92.08%. The product is a solid and appears white to off-white. It has a molecular weight of 131.17 and a formula of C6H13NO2.
  • Inhibits the growth of *Escherichia coli* HfrH
  • Chemosterilant and dietary additive
  • White to off-white solid appearance
Catalog No. 50-004-11907