Learn More
Carbosynth LLC. Leucine-Enkephalin (Human, Porcine, Bovine, Rat, Mouse), 25mg, MW: 555.62 • 18.02, Molecular formula: C28H37N5O7 • H2O, CAS# 58822-25-6
SDP
Supplier:  Carbosynth LLC. PEK404325MG
Encompass_Preferred
Synonyms: Tyr-Gly-Gly-Phe-Leu • H2O, Storage Temperature: -20°C, References: J. Hughes, T.W. Smith, H.W. Kosterlitz, L.A. Fothergill, B.A. Morgan, and H.R. Morris, Nature, 258, 577 (1975). (Original; Porcine), M. Comb, P.H. Seeburg, J. Adelman, L. Eiden, and E. Herbert, Nature, 295, 663 (1982). (cDNA Seq.; Human), M. Noda, Y. Furutani, H. Takahashi, M. Toyosato, T. Hirose, S. Inayama, S. Nakanishi, and S. Numa, Nature, 295, 202 (1982). (cDNA Seq.; Bovine)
Catalog No. 50-168-6459