Learn More
Medchemexpress LLC HY-19948 5mg , Leucomethylene blue (mesylate) CAS:1236208-20-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY199485MG
Encompass_Preferred
HY-19948 5mg Leucomethylene blue (mesylate) CAS: 1236208-20-0 Leucomethylene blue (TRx0237) mesylate, an orally active second-generation tau protein aggregation inhibitor (Ki of 0.12 μM), could be used for the study of Alzheimer's Disease. Leucomethylene blue mesylate is a common reduced form of Methylene Blue, Methylene Blue is a member of the thiazine class of dyes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1912