Learn More
Cayman Chemical LeucomycIn A4 5mg
SDP
Supplier:  Cayman Chemical 280665 MG
Encompass_Preferred
A macrolide antibiotic; active against a variety of bacteria, including S. aureus, B. subtilis, C. diphtheriae, N. gonorrhoeae, and H. influenzae (MICs = 0.15, 1.25, 0.15, 0.6, and 0.15 UG/ml, respectively)
Catalog No. 50-293-9808