Learn More
ATCC Leuconostoc; mesenteroides; subsp. mesenteroides
Supplier:  ATCC 10830
Encompass
Leuconostoc mesenteroides subsp. mesenteroides strain NRRL B-512 was isolated from slime on a root beer. This whole-genome sequenced strain is known to produce dextran for use as a plasmid extender and substitute for mucin.
Catalog No. 50-238-3297
 Restricted Item