Learn More
Medchemexpress LLC Leucyl-phenylalanine | 3063-05-6 | 99.6% | 278.35 | 1 ML

Supplier: Medchemexpress LLC HY11327810MM/1ML
Leucyl-phenylalanine is an endogenous metabolite classified as a dipeptide. This compound is strictly for research purposes only.
- Endogenous metabolite
- Classified as a dipeptide
- Purity of 99.56%
- Appears as a white to off-white solid
- Specific storage conditions for powder
- Specific storage conditions for solutions
- Soluble in water and ethanol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.