Learn More
Cayman Chemical LeukotrIen B4 100ug
SDP
Supplier:  Cayman Chemical 20110100 UG
Encompass_Preferred
A key dihydroxy fatty acid derived from AA through the 5-LO pathway; promotes a number of leukocyte functions including chemotaxis and chemokinesis at subnanomolar concentrations
Catalog No. 50-298-5167