Learn More
Cayman Chemical LeukotrIen B4-d4 25ug
SDP
Supplier:  Cayman Chemical 32011025 UG
Encompass_Preferred
An internal standard for the quantification of LTB4 by GC- or LC-MS.
Catalog No. 50-294-7014