Learn More
Cayman Chemical LeukotrIen B4 EthnolamID 100ug
SDP
Supplier:  Cayman Chemical 20112100 UG
Encompass_Preferred
A theoretical 5-LO metabolite of AEA; acts as a potent antagonist of the human BLT1 receptor (Ki = 1.22 nM)
Catalog No. 50-298-4520