Learn More
Cayman Chemical LeukotrIen C4 100ug
SDP
Supplier:  Cayman Chemical 20210100 UG
Encompass_Preferred
The parent CysLT produced by the LTC4 synthase-catalyzed conjugation of glutathione to LTA4; potent inducer of bronchoconstriction and enhanced vascular permeability that contributes to the pathogenesis of asthma and acute allergic hypersensitivity
Catalog No. 50-298-1035