Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Leukotriene A4 Hydrolase Antibody (YA2400) | 10 UL
SDP

Supplier:  Medchemexpress LLC HYP8265510UL

Encompass_Preferred

Leukotriene A4 Hydrolase Antibody (YA2400) is a rabbit-derived antibody for research use. It is supplied as a liquid at a concentration of 0.3 mg/mL and is stable under recommended storage conditions, specifically at -20°C.

  • Target: Leukotriene A4 Hydrolase (LTA4; LTA4 hydrolase; LTA4H)
  • Reactivity: Human, mouse, rat
  • Molecular weight: 69 kDa
  • Applications: WB, IHC-P, ICC/IF, FC
  • Formulation: 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide, 0.05% BSA
  • Storage: Store at -20°C for 1 year, avoid repeated freeze/thaw cycles.
  • For research use only.

Catalog No. 50-004-43119


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.