Learn More
Cayman Chemical Leukotriene A4 Hydrolase hum
SDP
Supplier:  Cayman Chemical 10007817100 UG
Encompass_Preferred
Source: Active recombinant C-terminal His-tagged enzyme expressed in E. coli • MW: ~69 kDa • A bifunctional zinc metalloenzyme that converts LTA4 into LTB4, a potent neutrophil chemoattractant; potential drug target for a variety of indications associated with leukocyte infiltration to sites of inflammation; has application for the screening of inhibitors of LTB4 synthesis
Catalog No. 50-288-9422