Learn More
Enzo Life Sciences Leukotriene B4 (1mg). CAS: 71160-24-2
SDP
Supplier:  Enzo Life Sciences BMLLB0041000
Encompass_Preferred
Activator of PPARα. Stimulates leukocyte functions including chemokinesis, chemotaxis, lysosomal enzyme release, adhesion, stimulation of ion fluxes, superoxyde anion production and C3b receptor expression. Induces vascular permeability, NK cell activity, and broncho-constriction. UVmax: 270 nm (50,000). Alternative name: LTB4, 5,12-Dihydroxy-[S-[R*,S*-(E,Z,Z,E)]]-6,8,10,14-eicosatetraenoic acid. Formulation: Oil dissolved in ethanol. Purity: ≥97% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-1143