Learn More
Medchemexpress LLC Leukotriene E4 | 75715-89-8 | 99.8% | 439.61 | 100 UG
Supplier: Medchemexpress LLC HY113465100UG
Leukotriene E4 is produced by the action of dipeptidase on LTD4. It is a constituent of slow-reacting substance of anaphylaxis (SRS-A). Leukotriene E4 accumulates in both plasma and urine, and its urinary excretion is often used as an indicator of asthma.