Learn More
Medchemexpress LLC Leukotriene E4 | 75715-89-8 | 99.8% | 439.61 | 25 UG

Supplier: Medchemexpress LLC 25UGHY113465
Leukotriene E4 (LTE4) is produced by the action of dipeptidase on LTD4. It is a constituent of slow-reacting substance of anaphylaxis (SRS-A) and accumulates in both plasma and urine. Urinary excretion of Leukotriene E4 is often used as an indicator of asthma.
- Appearance: Liquid
- Color: Colorless to light yellow
- Human endogenous metabolite
- Solution storage at -20°C for 2 years
- Ships at room temperature in continental US
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.