Learn More
Crescent Chemical Co Inc LEUPEPTIN
SDP
Supplier:  Crescent Chemical Co Inc SE51867.03
Encompass_Preferred
Leupeptin Inhibitor of serine and cysteine proteases. Size - 50MG Storage Conditions - -15 °C TO -25 °C Catalog Number - 51867.03 CAS 103476-89-7
Catalog No. 50-249-203