Learn More
Carbosynth LLC. Leupeptin, 25mg, MW: 426.55 49.04 , Molecular formula: C20H38N6O4 H2SO, CAS# 103476-89-7
SDP
Supplier:  Carbosynth LLC. ILP40410.025G
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Acetyl-L-Leucyl-L-Leucyl-L-Argininal Hemisulfate; Ac-LLR-CHO, Application: Microbial Product Inhibitor for Trypsin, Plasmin, Papain, and Cathepsin, Storage Temperature: -20C, References: S. Kondo, K. Kawamura, J. Iwanaga, M. Hamada, T. Aoyagi, K. Maeda, T. Takeuchi, and H. Umezawa, Chem. Pharm. Bull., 17, 1896 (1969). (Biological Activity), T. Aoyagi, T. Takeuchi, A. Matsuzaki, K. Kawamura, S. Kondo, M. Hamada, K. Maeda, and H. Umezawa, J. Antibiotics, 22, 283 (1969). (Original), T. Aoyagi, S. Miyata, M. Nanbo, F. Kojima, M. Matsuzaki, M. Ishizuka, T. Takeuchi, and H. Umezawa, J. Antibiotics, 22, 558 (1969). (Biological Activity)
Catalog No. 50-168-5877