Learn More
Carbosynth LLC. Leupeptin (0.5 mg vial), 1ea, MW: 426.55, Molecular formula: C20H38N6O4, CAS# 55123-66-5
SDP
Supplier:  Carbosynth LLC. ILP4041V0.5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Acetyl-L-Leucyl-L-Leucyl-L-Argininal; Ac-LLR-CHO, Application: Microbial Product Inhibitor for Trypsin, Plasmin, Papain, and Cathepsin, Salt Form:, Sulfate Form, Storage Temperature: -20C, References: S. Kondo, K. Kawamura, J. Iwanaga, M. Hamada, T. Aoyagi, K. Maeda, T. Takeuchi, and H. Umezawa, Chem. Pharm. Bull., 17, 1896 (1969). (Biological Activity), T. Aoyagi, T. Takeuchi, A. Matsuzaki, K. Kawamura, S. Kondo, M. Hamada, K. Maeda, and H. Umezawa, J. Antibiotics, 22, 283 (1969). (Original), T. Aoyagi, S. Miyata, M. Nanbo, F. Kojima, M. Matsuzaki, M. Ishizuka, T. Takeuchi, and H. Umezawa, J. Antibiotics, 22, 558 (1969). (Biological Activity)
Catalog No. 50-168-5878