Learn More
Carbosynth LLC. Leupeptin, 1g, MW: 426.55 • 49.04 , Molecular formula: C20H38N6O4 • ½H2SO, CAS# 103476-89-7
SDP
Supplier:  Carbosynth LLC. ILP40411G
Encompass_Preferred
Synonyms: Acetyl-L-Leucyl-L-Leucyl-L-Argininal Hemisulfate; Ac-LLR-CHO, Application: Microbial Product Inhibitor for Trypsin, Plasmin, Papain, and Cathepsin, Storage Temperature: -20°C, References: S. Kondo, K. Kawamura, J. Iwanaga, M. Hamada, T. Aoyagi, K. Maeda, T. Takeuchi, and H. Umezawa, Chem. Pharm. Bull., 17, 1896 (1969). (Biological Activity), T. Aoyagi, T. Takeuchi, A. Matsuzaki, K. Kawamura, S. Kondo, M. Hamada, K. Maeda, and H. Umezawa, J. Antibiotics, 22, 283 (1969). (Original), T. Aoyagi, S. Miyata, M. Nanbo, F. Kojima, M. Matsuzaki, M. Ishizuka, T. Takeuchi, and H. Umezawa, J. Antibiotics, 22, 558 (1969). (Biological Activity)
Catalog No. 50-168-5876