Learn More
Research Products International Corp Leupeptin Hemisulfate, 100 Milligrams
SDP
Supplier:  Research Products International Corp L220350.1
Encompass_Preferred
Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin.
Working concentration: 1-10uM
Catalog No. 50-136-8325