Learn More
Selleck Chemical LLC Leupeptin Hemisulfate
SDP
Supplier:  Selleck Chemical LLC S73801G
Encompass_Preferred
Leupeptin Hemisulfate is a reversible inhibitor of serine and cysteine proteases It inhibits cathepsin B (Ki 6 nM) calpain (Ki 10 nM) trypsin (Ki 35 nM) plasmin (Ki 3 4 M) and kallikrein (Ki 19 M) and has no effect against chymotrypsin elastase renin or pepsin
Catalog No. 50-000-04584