Learn More
Carbosynth LLC. Leupeptin (Synthetic), 25mg, MW: 426.55 49.04, Molecular formula: C20H38N6O4 H2SO4, CAS# 103476-89-7
SDP
Supplier:  Carbosynth LLC. FL2962825MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Ac-Leu-Leu-Arg-H H2SO4; Leupeptin Hemisulfate (Synthetic), Salt Form:, Hemisulfate salt, Storage Temperature: -20C
Catalog No. 50-168-5874