Learn More
Medchemexpress LLC Levacetylleucine | 1188-21-2 | C8H15NO3 | 10 G
SDP
Supplier:  Medchemexpress LLC HY5929110G
Encompass_Preferred
Levacetylleucine (N-acetyl-L-leucine) is an orally bioavailable and brain-penetrant compound, derived from the amino acid Leucine through acetylation. It is the active form of N-acetyl-leucine (NAL). This compound offers several benefits and features for research:
  • Attenuates neuronal death and neuroinflammation in the cortical tissue of mice.
  • Potentially improves lysosomal and metabolic dysfunction.
  • Improves compensation of postural symptoms after unilateral chemical labyrinthectomy (UL) in rats.
  • Shows promise for research in neurological manifestations of Niemann-Pick disease type C, traumatic brain injury, and neurodegeneration prevention.
Catalog No. 50-004-34401