Learn More
Medchemexpress LLC Levamisole | 14769-73-4 | 99.97% | C11H12N2S | 1 ML

Supplier: Medchemexpress LLC HYA010610MM/1ML
Levamisole is an anthelmintic agent with immunomodulatory properties. It acts as a positive allosteric modulator (PAM) for the α3β2 and α3β4 subtypes of nAChRs, demonstrating oral activity. It has been shown to prevent weight gain in mice fed a high-fat diet and is suitable for various research applications.
- Acts as a positive allosteric modulator for nAChRs
- Possesses anthelmintic properties
- Exhibits immunomodulatory effects
- Orally active
- Can be stored as powder at -20°C for 3 years or 4°C for 2 years
- When in solvent, store at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.