Learn More
Medchemexpress LLC Levamlodipine (besylate) | 150566-71-5 | 99.8% | 567.05 | 25 MG

Supplier: Medchemexpress LLC 25MGHY14744A
Levamlodipine besylate is an orally active calcium channel blocker that exhibits antioxidant and vasodilatory properties. It is utilized in research for vascular dementia, hypertension, and cerebrovascular diseases, capable of reducing serum malondialdehyde (MDA) levels and increasing superoxide dismutase (SOD) activity to improve oxidative stress.
- Appearance as a white to light yellow solid
- Stable for storage at -20°C, sealed, away from moisture and light
- Soluble in DMSO and H2O
- Targets calcium channels
- Reduces serum malondialdehyde (MDA) levels
- Increases superoxide dismutase (SOD) activity
- Improves oxidative stress
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.