Learn More
Grainger Level Mount Swivel Stud 1/2-13 1-7/8 in.
Supplier:  Grainger 14H403
Encompass_Preferred
The S&W Manufacturing Male End Leveling Mount Swiveling Pad is a leveling mount designed for RV and trailer applications. It features a 3" diameter swiveling pad that provides 360 degrees of rotation. The mount is constructed from galvanized steel and has a weight capacity of 3000 lbs. It comes with a 5/8" diameter threaded shank that fits most standard trailer frames. The height is adjustable from 13 3/4" min to 15 3/4" max. Overall, this is a heavy duty swiveling leveling mount ideal for stabilizing and leveling RVs, travel trailers, and cargo trailers.
Catalog No. 50-275-1172