Learn More
Grainger Level Mount Swivel Stud 5/8-11 2-1/2 in.
Supplier:  Grainger 14H405
Encompass_Preferred
The S-W Manufacturing Leveling Mount is a heavy duty leveling mount made of steel with a zinc plated finish. It has a weight capacity of 500 lbs and a minimum height of 5.75 inches and maximum height of 11 inches. The mount features a 5/8 inch diameter threaded post and has leveling feet that can adjust up to 3 inches for leveling. This sturdy mount provides a stable base for machinery, equipment, signs, gates, and more. With its rugged steel construction and wide height adjustment range, this leveling mount is ideal for industrial, commercial, and agricultural applications requiring secure, adjustable mounting.
Catalog No. 50-275-2307