Learn More
Grainger Level Mount Swivel Stud M10 1-1/4in Base
Supplier:  Grainger 14H414
Encompass_Preferred
The Level Mount Swivel Stud M10-1.5 with 1-1/4" Base from Grainger is a leveling mount with a swivel stud that allows equipment to be leveled and aligned. It is made from zinc-plated steel and has an M10 x 1.5 threaded stud with a swivel radius of 360 degrees. The mount has a wide 1-1/4" base that provides stability. This versatile leveling mount is suitable for leveling and aligning equipment like conveyors, pumps, motors and more. With its durable construction and swivel function, it's a useful leveling solution for industrial applications.
Catalog No. 50-275-2184