Learn More
Caron Products And Services Foot, Low Profile Leveling, 1/2"-13 Thread, with 2 Jam Nuts Included, Clear Zinc Finish, Steel
Supplier:  Caron Products And Services FTV109
Encompass_Preferred
Foot, Low Profile Leveling, 1/2"-13 Thread, with 2 Jam Nuts Included, Clear Zinc Finish, Steel
Catalog No. NC2402724