Learn More
Grainger Leveling Mount Swivel Stud 1-8 4 in Base
Supplier:  Grainger 14H410
Encompass_Preferred
The S-W Manufacturing Leveling Mount is a heavy duty leveling mount made of steel with a zinc dichromate finish. It has a weight capacity of 500 lbs and a minimum height of 1.8 inches. The leveling mount features a 5/8 inch diameter threaded stem and measures 2 inches wide x 2 inches long. It can adjust up to 1 inch for leveling and has a threaded hex nut with spanner holes for easy adjustment. The leveling mount is designed for secure, heavy duty mounting applications.
Catalog No. 50-275-2095