Learn More
Apexbio Technology LLC Levetiracetam 102767-28-2 100mg
SDP
Supplier:  Apexbio Technology LLC A1198100
Encompass_Preferred
Levetiracetam (CAS 102767-28-2) is an antiepileptic small-molecule agent recognized for its distinct mode of action compared to traditional anticonvulsants It binds selectively to synaptic vesicle glycoprotein 2A (SV2A) a protein involved in the regulation of intracellular calcium dynamics and neurotransmitter release Mechanistically levetiracetam negatively modulates gamma-aminobutyric acid (GABA)- and glycine-gated neuronal currents Importantly levetiracetam does not induce hepatic cytochrome P450 enzymes exhibits negligible plasma protein binding and is predominantly excreted unchanged renally with partial hydrolytic metabolism clinically levetiracetam is utilized widely in research addressing partial and generalized epileptic seizures
Catalog No. 50-000-62776