Learn More
U.S. Pharmacopeia Levetiracetam, 102767-28-2, MFCD03265610, 200 mg
Supplier:  U.S. Pharmacopeia 1359404
Encompass
Empirical Formula (Hill Notation): C8H14N2O2, Molecular Weight: 170.21, Synonym: (αS)-α-Ethyl-2-oxo-1-pyrrolidineacetamide, 2(S)-(2-Oxopyrrolidin-1-yl)butyramide.
Catalog No. NC1455586