Learn More
TARGETMOL CHEMICALS INC Levetiracetam 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0192200MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Levetiracetam (SIB-S1) is a relatively unique anticonvulsant that is typically used in combination with other antiepileptic medications for partial onset seizures. Levetiracetam has been linked to rare instances of serum aminotransferase and alkaline phosphatase elevations during treatment and to rare cases of clinically apparent drug induced liver disease. Purity 99.81%
Catalog No. 50-288-0564