Learn More
Selleck Chemical LLC Levobetaxolol HCl S4085-50mg
SDP
Supplier:  Selleck Chemical LLC S408550MG
Encompass_Preferred
Levobetaxolol (AL 1577A (S)-Betaxolol hydrochloride) exhibits a higher affinity at cloned human 1 and 2 receptors with Ki value of 0 76 nM and 32 6 nM respectively
Catalog No. 50-310-9498