Learn More
Selleck Chemical LLC Levobunolol E2875-25MG
SDP
Supplier:  Selleck Chemical LLC E287525MG
Encompass_Preferred
Levobunolol (l-Bunolol) hydrochloride is a potent and nonselective -adrenergic receptor antagonist with an IC50 of 48 M
Catalog No. 50-000-40697