Learn More
Medchemexpress LLC Levobunolol (hydrochloride) | 27912-14-7 | MFCD00864586 | 100.0% | 5 MG
Supplier: Medchemexpress LLC HYB10355MG
Levobunolol hydrochloride is a non-selective β-adrenergic antagonist and vasodilator. It is widely used in glaucoma and ocular hypertension research due to its ability to dilate ciliary arteries, increase ocular blood flow, and block calcium ion influx, which reduces vascular smooth muscle sensitivity to calcium.
- Inhibits β-receptor signaling pathway.
- Reduces proliferation markers in corneal cells.
- Accelerates healing of mechanical injury.
- Inhibits histamine-induced vasoconstriction.
- Protects ocular blood flow and promotes corneal repair.