Learn More
Cayman Chemical LevobupIvacaIn-d9hydrochlo 5mg
SDP
Supplier:  Cayman Chemical 389045 MG
Encompass_Preferred
An internal standard for the quantification of levobupivacaine by GC- or LC-MS
Catalog No. 50-291-8873