Learn More
Cayman Chemical LevobupIvacaInd9hydrochlo 10mg
SDP
Supplier:  Cayman Chemical 3890410 MG
Encompass_Preferred
An internal standard for the quantification of levobupivacaine by GC- or LC-MS
Catalog No. 50-291-8910