Learn More
Cayman Chemical LevocabastInhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 2101410 MG
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 62.5 nM); selective for histamine H1 over H2 and H3 receptors (Kis = 23.5 and 1.94 µM, respectively); binds to the rat neurotensin-2 receptor (IC50 = 10 nM); reduces the severity of ovalbumin-induced conjunctivitis in ovalbumin-sensitized rats when applied ocularly to ovalbumin-sensitized rats
Catalog No. 50-298-2714