Learn More
Medchemexpress LLC HY-W010841 10mg Medchemexpress, Levocetirizine (dihydrochloride) CAS:130018-87-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW01084110MG
Encompass_Preferred
Medchemexpress, HY-W010841 10mg Levocetirizine (diHCl) CAS:130018-87-0 Levocetirizine dihydrochloride ((R)-Cetirizine dihydrochloride) is a third-generation peripheral H1-receptor antagonist. Levocetirizine dihydrochloride is an antihistaminic agent which is the R-enantiomer of Cetirizine. Levocetirizine dihydrochloride has a higher affinity for the histamine H1-receptor than (S)-Cetirizine and can effectively treat allergic rhinitis and chronic idiopathic urticaria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0679