Learn More
Medchemexpress LLC Levocetirizine dihydrochloride | 130018-87-0 | 99.8% | 461.81 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW010841500MG
Encompass_Preferred
Levocetirizine dihydrochloride is a third-generation peripheral H1-receptor antagonist. It is an antihistaminic agent, specifically the R-enantiomer of Cetirizine. This compound demonstrates a higher affinity for the histamine H1-receptor compared to (S)-Cetirizine, making it effective in treating allergic rhinitis and chronic idiopathic urticaria. It is intended for research use only.
  • Third-generation peripheral H1-receptor antagonist
  • R-enantiomer of Cetirizine
  • Higher affinity for the histamine H1-receptor compared to (S)-Cetirizine
  • Effective in treating allergic rhinitis and chronic idiopathic urticaria
  • Intended for research use only
Catalog No. 50-003-66503